Assay Details
Target Gene Details
Entrez Gene ID: | 25842 |
Gene Name: | anti-silencing function 1A histone chaperone |
Gene Aliases: |
CGI-98, CIA, HSPC146 |
Location: |
Chr.6:118894077-118909171 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASF1A | NM_014034.2 | NP_054753.1 | ||
| AB028628.1 | BAA96542.1 | |||
| AF151856.1 | AAD34093.1 | |||
| AF161495.1 | AAF29110.1 | |||
| AF279306.1 | AAK82972.1 | |||
| AK025738.1 | BAB15228.1 | |||
| AL050261.1 | CAB43363.1 | |||
| BC010878.1 | AAH10878.1 | |||
| CR457347.1 | CAG33628.1 | |||
| DA902777.1 |
Target Gene Details
Entrez Gene ID: | 254394 |
Gene Name: | minichromosome maintenance 9 homologous recombination repair factor |
Gene Aliases: |
C6orf61, MCMDC1, ODG4, dJ329L24.1, dJ329L24.3 |
Location: |
Chr.6:118813449-118935162 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM9 | NM_017696.2 | NP_060166.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161446 | Chr.6:118418441 - 118902959 on Build GRCh38 | Duplication |
|
| nsv509151 | Chr.6:118823819 - 118901881 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map