Assay Details
Target Gene Details
Entrez Gene ID: | 2898 |
Gene Name: | glutamate ionotropic receptor kainate type subunit 2 |
Gene Aliases: |
EAA4, GLR6, GLUK6, GLUR6, GluK2, MRT6 |
Location: |
Chr.6:101393708-102070083 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIK2 | NM_001166247.1 | NP_001159719.1 | ||
| NM_021956.4 | NP_068775.1 | |||
| NM_175768.3 | NP_786944.1 | |||
| XM_005266945.2 | XP_005267002.1 | |||
| XM_011535777.2 | XP_011534079.1 | |||
| XM_011535778.2 | XP_011534080.1 | |||
| XM_017010781.1 | XP_016866270.1 | |||
| XM_017010782.1 | XP_016866271.1 | |||
| AJ252246.1 | CAC81020.1 | |||
| AJ301610.1 | CAC67487.1 | |||
| AK289542.1 | ||||
| AK291965.1 | ||||
| BC037954.1 | AAH37954.1 | |||
| BC063814.1 | AAH63814.1 | |||
| BQ717946.1 | ||||
| HM149335.1 | ||||
| HM149336.1 | ||||
| HM149337.1 | ||||
| HM149338.1 | ||||
| HM149339.1 | ||||
| U16126.1 | AAC50420.1 |
More Information
Set Membership: |
|

Genomic Map