Assay Details
Target Gene Details
Entrez Gene ID: | 221393 |
Gene Name: | adhesion G protein-coupled receptor F4 |
Gene Aliases: |
GPR115, PGR18 |
Location: |
Chr.6:47698553-47722021 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRF4 | NM_153838.3 | NP_722580.3 | ||
| AK095395.1 | ||||
| DA445316.1 | ||||
| DQ315370.1 | ABC41929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017113 | Chr.6:46846140 - 48020279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map