Assay Details
Target Gene Details
Entrez Gene ID: | 134728 |
Gene Name: | interleukin 1 receptor associated kinase 1 binding protein 1 |
Gene Aliases: |
AIP70, SIMPL |
Location: |
Chr.6:78867486-78979411 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IRAK1BP1 | AK309003.1 |
Target Gene Details
Entrez Gene ID: | 55023 |
Gene Name: | pleckstrin homology domain interacting protein |
Gene Aliases: |
BRWD2, DCAF14, WDR11, ndrp |
Location: |
Chr.6:78934419-79078294 on Build GRCh38 |
Assay Gene Location: | Within Exon 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHIP | NM_017934.5 | 40 | 6998 | NP_060404.3 |
More Information
Additional Information:
For this assay, SNP(s) [rs145326834] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map