Assay Details
Target Gene Details
Entrez Gene ID: | 313 |
Gene Name: | acyloxyacyl hydrolase |
Gene Aliases: |
- |
Location: |
Chr.7:36512042-36724549 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Gene Details
Entrez Gene ID: | 100874264 |
Gene Name: | AOAH intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.7:36597834-36600120 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AOAH-IT1 | NR_046764.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464427 | Chr.7:36577662 - 36792118 on Build GRCh38 | Loss |
|
| nsv830958 | Chr.7:36572305 - 36731903 on Build GRCh38 | Gain |
|
| nsv830957 | Chr.7:36464021 - 36636017 on Build GRCh38 | Gain |
|
| nsv1032157 | Chr.7:36591870 - 36627946 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map