Assay Details
Target Gene Details
Entrez Gene ID: | 50640 |
Gene Name: | patatin like phospholipase domain containing 8 |
Gene Aliases: |
IPLA2-2, IPLA2G, MMLA, PNPLA-gamma, iPLA2gamma |
Location: |
Chr.7:108470417-108528161 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNPLA8 | NM_001256011.2 | NP_001242940.1 | ||
| XM_005250396.4 | 1 | 749 | XP_005250453.1 | |
| XM_011516274.2 | 1 | 749 | XP_011514576.1 | |
| XM_011516275.2 | 1 | 749 | XP_011514577.1 | |
| XM_017012261.1 | 1 | 749 | XP_016867750.1 | |
| AI742208.1 | 1 | 107 | ||
| AK024335.1 | BAB14890.1 |
More Information
Set Membership: |
|

Genomic Map