Assay Details
Target Gene Details
Entrez Gene ID: | 55972 |
Gene Name: | solute carrier family 25 member 40 |
Gene Aliases: |
MCFP |
Location: |
Chr.7:87834499-87876377 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A40 | NM_018843.3 | NP_061331.2 | ||
| AF125531.1 | AAF17225.1 | |||
| AK223020.1 | BAD96740.1 | |||
| AK290848.1 | ||||
| BC027322.1 | AAH27322.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607725 | Chr.7:87203287 - 87911933 on Build GRCh38 | Gain |
|
| esv2734777 | Chr.7:87840405 - 87848775 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map