Assay Details
Target Gene Details
Entrez Gene ID: | 26047 |
Gene Name: | contactin associated protein-like 2 |
Gene Aliases: |
AUTS15, CASPR2, CDFE, NRXN4, PTHSL1 |
Location: |
Chr.7:146116035-148420998 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTNAP2 | NM_014141.5 | NP_054860.1 | ||
| AB020675.1 | ||||
| AB073883.1 | ||||
| AF319045.1 | AAK34932.1 | |||
| AK296532.1 | ||||
| AK315113.1 | ||||
| BC093780.1 | AAH93780.1 | |||
| BC113373.1 | ||||
| KJ534811.1 | ||||
| KJ535088.1 |
Target Gene Details
Entrez Gene ID: | 105375556 |
Gene Name: | uncharacterized LOC105375556 |
Gene Aliases: |
- |
Location: |
Chr.7:147671679-147673144 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105375556 | NR_133929.1 | 2 | 971 | |
| NR_133930.1 | 2 | 614 | ||
| BF188822.1 | 2 | 956 | ||
| BF188823.1 | 1 | 209 | ||
| BX385984.2 | 1 | 133 | ||
| BX385985.2 | 2 | 614 |
More Information
Set Membership: |
|

Genomic Map