Assay Details
Target Gene Details
Entrez Gene ID: | 101927870 |
Gene Name: | uncharacterized LOC101927870 |
Gene Aliases: |
- |
Location: |
Chr.7:103445207-103514007 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927870 | NR_110141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576411 | Chr.7:103468275 - 103474748 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73712385] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map