Assay Details
Target Gene Details
Entrez Gene ID: | 168667 |
Gene Name: | BMP binding endothelial regulator |
Gene Aliases: |
CRIM3, CV-2, CV2 |
Location: |
Chr.7:33904911-34155872 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BMPER | NM_133468.4 | NP_597725.1 | ||
| XM_005249633.2 | XP_005249690.1 | |||
| XM_017011800.1 | XP_016867289.1 | |||
| XM_017011801.1 | XP_016867290.1 | |||
| AK289963.1 | ||||
| AY324650.1 | AAP89012.1 | |||
| BC060868.1 | AAH60868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1080e201 | Chr.7:32621750 - 35008557 on Build GRCh38 | Deletion |
|
| esv3612779 | Chr.7:33861781 - 33942872 on Build GRCh38 | Gain |
|
| esv3612772 | Chr.7:33730657 - 34432276 on Build GRCh38 | Loss |
|
| nsv1024493 | Chr.7:33733357 - 34425137 on Build GRCh38 | Loss |
|
| nsv1135982 | Chr.7:32794601 - 34893593 on Build GRCh38 | Deletion |
|
| nsv436907 | Chr.7:32668474 - 34999437 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map