Assay Details
Target Gene Details
Entrez Gene ID: | 313 |
Gene Name: | acyloxyacyl hydrolase |
Gene Aliases: |
- |
Location: |
Chr.7:36512042-36724549 on Build GRCh38 |
Assay Gene Location: | Within Intron 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AOAH | NM_001177506.1 | NP_001170977.1 | ||
| NM_001177507.1 | NP_001170978.1 | |||
| NM_001637.3 | NP_001628.1 | |||
| XM_011515333.2 | XP_011513635.1 | |||
| XM_011515334.2 | XP_011513636.1 | |||
| XM_011515335.2 | XP_011513637.1 | |||
| XM_011515336.2 | XP_011513638.1 | |||
| XM_011515338.2 | XP_011513640.1 | |||
| XM_011515342.2 | XP_011513644.1 | |||
| XM_017012102.1 | XP_016867591.1 | |||
| XM_017012103.1 | XP_016867592.1 | |||
| XM_017012104.1 | XP_016867593.1 | |||
| XM_017012105.1 | XP_016867594.1 | |||
| AK223476.1 | BAD97196.1 | |||
| AK297016.1 | ||||
| AK298644.1 | ||||
| AK308215.1 | ||||
| BC025698.1 | AAH25698.1 | |||
| CR748867.1 | ||||
| M62840.1 | AAA35506.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951630 | Chr.7:36514895 - 36516594 on Build GRCh38 | Deletion |
|
| nsv830957 | Chr.7:36464021 - 36636017 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map