Assay Details
Target Gene Details
Entrez Gene ID: | 4233 |
Gene Name: | MET proto-oncogene, receptor tyrosine kinase |
Gene Aliases: |
AUTS9, DFNB97, HGFR, RCCP2, c-Met |
Location: |
Chr.7:116672359-116798386 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MET | NM_000245.3 | NP_000236.2 | ||
| NM_001127500.2 | NP_001120972.1 | |||
| NM_001324401.1 | NP_001311330.1 | |||
| NM_001324402.1 | NP_001311331.1 | |||
| XM_011516223.1 | XP_011514525.1 | |||
| AK309532.1 | ||||
| BC130420.1 | ||||
| DB223186.1 | ||||
| DQ656040.2 | ||||
| EU176015.1 | ||||
| J02958.1 | AAA59591.1 | |||
| X54559.1 | CAB56793.1 |
More Information
Set Membership: |
|

Genomic Map