Assay Details
Target Gene Details
Entrez Gene ID: | 5506 |
Gene Name: | protein phosphatase 1 regulatory subunit 3A |
Gene Aliases: |
GM, PP1G, PPP1R3 |
Location: |
Chr.7:113876827-113919094 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R3A | NM_002711.3 | NP_002702.2 | ||
| XM_005250473.3 | XP_005250530.1 | |||
| AF024579.1 | AAB94597.1 | |||
| BC126451.1 | ||||
| BC126453.1 | ||||
| X78578.1 | CAA55316.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831100 | Chr.7:113821021 - 113977373 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map