Assay Details
Target Gene Details
Entrez Gene ID: | 673 |
Gene Name: | B-Raf proto-oncogene, serine/threonine kinase |
Gene Aliases: |
B-RAF1, B-raf, BRAF1, NS7, RAFB1 |
Location: |
Chr.7:140719331-140924764 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRAF | XM_005250045.2 | 19 | 8904 | XP_005250102.1 |
| XM_017012558.1 | 20 | 9024 | XP_016868047.1 | |
| XM_017012559.1 | 19 | 8870 | XP_016868048.1 | |
| AK094023.1 | 1 | 1527 | ||
| AK098095.1 | 1 | 2186 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015269 | Chr.7:140712664 - 140759021 on Build GRCh38 | Gain |
|
| nsv831157 | Chr.7:140541539 - 140727076 on Build GRCh38 | Gain |
|
| nsv831158 | Chr.7:140692422 - 140902515 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map