Assay Details
Target Gene Details
Entrez Gene ID: | 313 |
Gene Name: | acyloxyacyl hydrolase |
Gene Aliases: |
- |
Location: |
Chr.7:36512042-36724549 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AOAH | XM_011515333.2 | 21 | 2517 | XP_011513635.1 |
| XM_011515334.2 | 21 | 2546 | XP_011513636.1 | |
| XM_011515335.2 | 23 | 2656 | XP_011513637.1 | |
| XM_011515342.2 | 12 | 1547 | XP_011513644.1 | |
| XM_017012102.1 | 20 | 2398 | XP_016867591.1 | |
| XM_017012103.1 | 19 | 1923 | XP_016867592.1 | |
| XM_017012105.1 | 13 | 1707 | XP_016867594.1 | |
| AK298644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830957 | Chr.7:36464021 - 36636017 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map