Assay Details
Target Gene Details
Entrez Gene ID: | 83605 |
Gene Name: | CCM2 scaffolding protein |
Gene Aliases: |
C7orf22, OSM, PP10187 |
Location: |
Chr.7:44999746-45076470 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCM2 | NM_001167934.1 | NP_001161406.1 | ||
| NM_001167935.1 | NP_001161407.1 | |||
| NM_031443.3 | NP_113631.1 | |||
| NR_030770.1 | ||||
| XM_011515562.1 | XP_011513864.1 | |||
| XM_011515564.2 | XP_011513866.1 | |||
| XM_017012673.1 | XP_016868162.1 | |||
| AF370392.1 | AAQ15228.1 | |||
| BC004903.2 | AAH04903.1 | |||
| BC008859.2 | AAH08859.1 | |||
| BC016832.2 | AAH16832.1 | |||
| BC063663.1 | AAH63663.1 | |||
| DA420491.1 |
More Information
Set Membership: |
|

Genomic Map