Assay Details
Target Gene Details
Entrez Gene ID: | 8935 |
Gene Name: | src kinase associated phosphoprotein 2 |
Gene Aliases: |
PRAP, RA70, SAPS, SCAP2, SKAP-HOM, SKAP55R |
Location: |
Chr.7:26654772-26864743 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SKAP2 | XM_017012771.1 | XP_016868260.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830929 | Chr.7:26548269 - 26710116 on Build GRCh38 | Gain |
|
| nsv1020378 | Chr.7:26640261 - 26694253 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map