Assay Details
Target Gene Details
Entrez Gene ID: | 55744 |
Gene Name: | cytochrome c oxidase assembly factor 1 homolog |
Gene Aliases: |
C7orf44, MITRAC15 |
Location: |
Chr.7:43608452-43729541 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3612996 | Chr.7:43537785 - 43656356 on Build GRCh38 | Gain |
|
| esv997089 | Chr.7:43630014 - 43641441 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map