Assay Details
Target Gene Details
Entrez Gene ID: | 117 |
Gene Name: | ADCYAP receptor type I |
Gene Aliases: |
PAC1, PAC1R, PACAPR, PACAPRI |
Location: |
Chr.7:31052341-31114005 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCYAP1R1 | NM_001118.4 | NP_001109.2 | ||
| NM_001199635.1 | NP_001186564.1 | |||
| NM_001199636.1 | NP_001186565.1 | |||
| NM_001199637.1 | NP_001186566.1 | |||
| XM_005249618.4 | XP_005249675.1 | |||
| XM_006715645.2 | XP_006715708.1 | |||
| XM_017011736.1 | XP_016867225.1 | |||
| XM_017011737.1 | XP_016867226.1 | |||
| XM_017011738.1 | XP_016867227.1 | |||
| AI815550.1 | ||||
| AK226138.1 | ||||
| AK290046.1 | ||||
| DA809763.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6316n100 | Chr.7:30609596 - 31145482 on Build GRCh38 | Gain |
|
| esv2761316 | Chr.7:30615466 - 31127062 on Build GRCh38 | Gain |
|
| nsv527038 | Chr.7:30617851 - 31122331 on Build GRCh38 | Gain |
|
| nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion |
|
| nsv464418 | Chr.7:31032771 - 31076553 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map