Assay Details
Target Gene Details
Entrez Gene ID: | 673 |
Gene Name: | B-Raf proto-oncogene, serine/threonine kinase |
Gene Aliases: |
B-RAF1, B-raf, BRAF1, NS7, RAFB1 |
Location: |
Chr.7:140719331-140924764 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRAF | NM_004333.4 | NP_004324.2 | ||
| XM_005250045.2 | XP_005250102.1 | |||
| XM_017012558.1 | XP_016868047.1 | |||
| XM_017012559.1 | XP_016868048.1 | |||
| AK314346.1 | ||||
| BC101757.1 | AAI01758.1 | |||
| BC112079.1 | AAI12080.1 | |||
| BM263585.1 | ||||
| M21001.1 | AAA96495.1 | |||
| M95712.2 | AAA35609.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015269 | Chr.7:140712664 - 140759021 on Build GRCh38 | Gain |
|
| nsv831158 | Chr.7:140692422 - 140902515 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map