Assay Details
Target Gene Details
Entrez Gene ID: | 222194 |
Gene Name: | round spermatid basic protein 1 like |
Gene Aliases: |
- |
Location: |
Chr.7:77696426-77779803 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSBN1L | NM_198467.2 | NP_940869.2 | ||
| BC059402.1 | AAH59402.1 |

Genomic Map