Assay Details
Target Gene Details
Entrez Gene ID: | 221927 |
Gene Name: | BRCA1 associated ATM activator 1 |
Gene Aliases: |
BAAT1, C7orf27, RMFSL |
Location: |
Chr.7:2537810-2555758 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRAT1 | NM_152743.3 | 1 | 95 | NP_689956.2 |
| XM_005249643.4 | 1 | 57 | XP_005249700.1 | |
| XM_011515177.2 | 1 | 62 | XP_011513479.1 | |
| XM_011515179.2 | 1 | 56 | XP_011513481.1 | |
| XM_011515181.2 | 1 | 53 | XP_011513483.1 | |
| XM_011515183.1 | 1 | 90 | XP_011513485.1 | |
| XM_011515186.2 | 1 | 54 | XP_011513488.1 | |
| XM_017011834.1 | 1 | 52 | XP_016867323.1 | |
| XM_017011835.1 | 1 | 90 | XP_016867324.1 | |
| XM_017011836.1 | 1 | 51 | XP_016867325.1 | |
| AK131083.1 | BAC85133.1 | |||
| BP363179.1 | 1 | 95 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830887 | Chr.7:2507249 - 2621396 on Build GRCh38 | Loss |
|
| nsv605862 | Chr.7:2516523 - 2604885 on Build GRCh38 | Loss |
|
| nsv1026756 | Chr.7:2550749 - 2579836 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map