Assay Details
Target Gene Details
Entrez Gene ID: | 2918 |
Gene Name: | glutamate metabotropic receptor 8 |
Gene Aliases: |
GLUR8, GPRC1H, MGLUR8, mGlu8 |
Location: |
Chr.7:126438598-127253294 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM8 | NM_000845.2 | NP_000836.2 | ||
| NM_001127323.1 | NP_001120795.1 | |||
| NR_028041.1 | ||||
| XM_006715938.3 | XP_006716001.1 | |||
| XM_011516091.1 | XP_011514393.1 | |||
| XM_011516092.2 | XP_011514394.1 | |||
| XM_017012074.1 | XP_016867563.1 | |||
| AJ236921.1 | CAB36968.1 | |||
| AJ236922.1 | CAB36969.1 | |||
| AK290197.1 | ||||
| AK315203.1 | ||||
| AY608335.1 | AAT37959.1 | |||
| BC093725.1 | AAH93725.1 | |||
| BC101675.1 | AAI01676.1 | |||
| EU432125.1 | ||||
| U92459.1 | AAB51764.1 | |||
| U95025.1 | AAB72040.1 |
Target Gene Details
Entrez Gene ID: | 101928333 |
Gene Name: | uncharacterized LOC101928333 |
Gene Aliases: |
- |
Location: |
Chr.7:127215127-127229921 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928333 | NR_110194.1 | |||
| NR_110195.1 | ||||
| BQ438530.1 |
More Information
Set Membership: |
|

Genomic Map