Assay Details
Target Gene Details
Entrez Gene ID: | 102725541 |
Gene Name: | EGFR long non-coding downstream RNA |
Gene Aliases: |
Fabl, LINC01156 |
Location: |
Chr.7:55238436-55255635 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELDR | NR_110426.1 | |||
| DB084610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734425 | Chr.7:53868335 - 55917254 on Build GRCh38 | Deletion |
|
| nsv607032 | Chr.7:55216561 - 55241732 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116381092,rs117628015] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map