Assay Details
Target Gene Details
Entrez Gene ID: | 11281 |
Gene Name: | POU class 6 homeobox 2 |
Gene Aliases: |
RPF-1, WT5, WTSL |
Location: |
Chr.7:38977878-39468601 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POU6F2 | NM_001166018.1 | NP_001159490.1 | ||
| NM_007252.3 | NP_009183.3 | |||
| XM_011515113.2 | XP_011513415.1 | |||
| DN991890.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1024142 | Chr.7:38939128 - 39260845 on Build GRCh38 | Gain |
|
| nsv830967 | Chr.7:38874237 - 39029653 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map