Assay Details
Target Gene Details
Entrez Gene ID: | 223075 |
Gene Name: | coiled-coil domain containing 129 |
Gene Aliases: |
- |
Location: |
Chr.7:31514071-31658720 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC129 | NM_001257967.1 | NP_001244896.1 | ||
| NR_047565.1 | ||||
| XM_011515213.1 | XP_011513515.1 | |||
| XM_017011871.1 | XP_016867360.1 | |||
| XM_017011872.1 | XP_016867361.1 | |||
| AK128026.1 | BAC87235.1 | |||
| AK304387.1 | ||||
| BC132719.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891094 | Chr.7:31500343 - 31591980 on Build GRCh38 | Gain |
|
| nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map