Assay Details
Target Gene Details
Entrez Gene ID: | 222194 |
Gene Name: | round spermatid basic protein 1 like |
Gene Aliases: |
- |
Location: |
Chr.7:77696426-77779803 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSBN1L | NM_198467.2 | NP_940869.2 | ||
| BC046353.1 | AAH46353.1 | |||
| BC059402.1 | AAH59402.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831039 | Chr.7:77564611 - 77722887 on Build GRCh38 | Gain |
|

Genomic Map