Assay Details
Target Gene Details
Entrez Gene ID: | 100505854 |
Gene Name: | Alu-mediated CDKN1A/p21 transcriptional regulator (non-protein coding) |
Gene Aliases: |
RSBN1L-AS1 |
Location: |
Chr.7:77657660-77697345 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APTR | NR_038361.1 | |||
| NR_134251.1 | ||||
| NR_134253.1 | ||||
| NR_134254.1 | ||||
| AF290475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2657737 | Chr.7:77684805 - 77692604 on Build GRCh38 | Deletion |
|
| esv2734703 | Chr.7:77690199 - 77692971 on Build GRCh38 | Deletion |
|
| esv3613805 | Chr.7:77684823 - 77692571 on Build GRCh38 | Loss |
|
| nsv831039 | Chr.7:77564611 - 77722887 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117056050] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map