Assay Details
Target Gene Details
Entrez Gene ID: | 7328 |
Gene Name: | ubiquitin conjugating enzyme E2 H |
Gene Aliases: |
E2-20K, GID3, UBC8, UBCH, UBCH2 |
Location: |
Chr.7:129830732-129952960 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2H | NM_001202498.1 | 7 | 4553 | NP_001189427.1 |
| NM_003344.3 | 7 | 4949 | NP_003335.1 | |
| NM_182697.2 | 5 | 4856 | NP_874356.1 | |
| AK024519.1 | 1 | 1511 | ||
| AK026094.1 | 1 | 1634 | ||
| AL832212.1 | 1 | 4969 | ||
| BC006277.2 | AAH06277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3572138 | Chr.7:129818320 - 129831214 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map