Assay Details
Target Gene Details
Entrez Gene ID: | 10466 |
Gene Name: | component of oligomeric golgi complex 5 |
Gene Aliases: |
CDG2I, GOLTC1, GTC90 |
Location: |
Chr.7:107201743-107564514 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COG5 | NM_001161520.1 | NP_001154992.1 | ||
| NM_006348.3 | NP_006339.3 | |||
| NM_181733.2 | NP_859422.2 | |||
| AF058718.1 | AAC69276.1 | |||
| AK022835.1 | ||||
| AK314270.1 | ||||
| BC068540.1 | AAH68540.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114610933] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map