Assay Details
Target Gene Details
Entrez Gene ID: | 101927811 |
Gene Name: | uncharacterized LOC101927811 |
Gene Aliases: |
- |
Location: |
Chr.7:20328301-20331762 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927811 | NR_110119.1 | |||
| NR_110120.1 | ||||
| AV707592.1 | ||||
| HY150107.1 |

Genomic Map