Assay Details
Target Gene Details
Entrez Gene ID: | 3696 |
Gene Name: | integrin subunit beta 8 |
Gene Aliases: |
- |
Location: |
Chr.7:20329819-20415759 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB8 | XM_011515393.2 | 1 | 241 | XP_011513695.1 |
| XM_017012179.1 | 1 | 240 | XP_016867668.1 |
Target Gene Details
Entrez Gene ID: | 101927811 |
Gene Name: | uncharacterized LOC101927811 |
Gene Aliases: |
- |
Location: |
Chr.7:20328301-20331762 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927811 | NR_110119.1 | |||
| NR_110120.1 | ||||
| AV707592.1 | ||||
| HY150107.1 |
More Information
Set Membership: |
|

Genomic Map