Assay Details
Target Gene Details
Entrez Gene ID: | 9586 |
Gene Name: | cAMP responsive element binding protein 5 |
Gene Aliases: |
CRE-BPA, CREB-5 |
Location: |
Chr.7:28299321-28825894 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB5 | NM_182899.4 | NP_878902.2 | ||
| L05913.1 | AAC37526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830931 | Chr.7:28132064 - 28329752 on Build GRCh38 | Gain |
|
| nsv5678 | Chr.7:28280351 - 28325206 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73291256] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map