Assay Details
Target Gene Details
Entrez Gene ID: | 5294 |
Gene Name: | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma |
Gene Aliases: |
PI3CG, PI3K, PI3Kgamma, PIK3, p110gamma, p120-PI3K |
Location: |
Chr.7:106865278-106908978 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3CG | NM_001282426.1 | NP_001269355.1 | ||
| NM_001282427.1 | NP_001269356.1 | |||
| NM_002649.3 | NP_002640.2 | |||
| XM_005250443.3 | XP_005250500.1 | |||
| XM_011516316.1 | XP_011514618.1 | |||
| XM_011516317.1 | XP_011514619.1 | |||
| XM_017012328.1 | XP_016867817.1 | |||
| AF327656.1 | AAG61115.1 | |||
| AK292684.1 | ||||
| AK313753.1 | ||||
| BC035683.1 | AAH35683.1 | |||
| BX648341.1 | ||||
| X83368.1 | CAA58284.1 |
More Information
Set Membership: |
|

Genomic Map