Assay Details
Target Gene Details
Entrez Gene ID: | 5683 |
Gene Name: | proteasome subunit alpha 2 |
Gene Aliases: |
HC3, MU, PMSA2, PSC2 |
Location: |
Chr.7:42916861-42932206 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMA2 | NM_002787.4 | NP_002778.1 | ||
| AK222976.1 | BAD96696.1 | |||
| AK290654.1 | ||||
| AU120930.1 | ||||
| BC002900.2 | AAH02900.2 | |||
| BC047697.1 | AAH47697.1 | |||
| BX641097.1 | 5 | 3867 | CAE46046.1 | |
| CB990982.1 | ||||
| CR450317.1 | CAG29313.1 | |||
| D00760.1 | BAA00657.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv606715 | Chr.7:42169735 - 42940343 on Build GRCh38 | Gain |
|
| nsv830979 | Chr.7:42806983 - 42987012 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map