Assay Details
Target Gene Details
Entrez Gene ID: | 51667 |
Gene Name: | negative regulator of ubiquitin like proteins 1 |
Gene Aliases: |
BS4, NUB1L, NYREN18 |
Location: |
Chr.7:151341699-151378461 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUB1 | NM_001243351.1 | NP_001230280.1 | ||
| NM_016118.4 | NP_057202.3 | |||
| XM_005250011.2 | XP_005250068.1 | |||
| XM_017012304.1 | XP_016867793.1 | |||
| XM_017012305.1 | XP_016867794.1 | |||
| XM_017012306.1 | XP_016867795.1 | |||
| XM_017012307.1 | XP_016867796.1 | |||
| XM_017012308.1 | XP_016867797.1 | |||
| XM_017012309.1 | XP_016867798.1 | |||
| XM_017012310.1 | XP_016867799.1 | |||
| XM_017012311.1 | XP_016867800.1 | |||
| XM_017012312.1 | XP_016867801.1 | |||
| XM_017012313.1 | XP_016867802.1 | |||
| XM_017012314.1 | XP_016867803.1 | |||
| AF155099.1 | AAD42865.1 | |||
| AF300717.1 | AAK21001.1 | |||
| AF459743.1 | AAO14547.1 | |||
| AK026433.1 | 5 | 871 | ||
| AY129295.1 | AAN01355.1 | |||
| BC046354.2 | AAH46354.1 | |||
| BG105733.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3615478 | Chr.7:151282115 - 151376312 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map