Assay Details
Target Gene Details
Entrez Gene ID: | 644 |
Gene Name: | biliverdin reductase A |
Gene Aliases: |
BLVR, BVR, BVRA |
Location: |
Chr.7:43758153-43807342 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLVRA | NM_000712.3 | NP_000703.2 | ||
| NM_001253823.1 | NP_001240752.1 | |||
| XM_011515474.2 | XP_011513776.1 | |||
| XM_017012520.1 | XP_016868009.1 | |||
| AK291862.1 | ||||
| BC005902.1 | AAH05902.1 | |||
| BC008456.1 | AAH08456.1 | |||
| BM710258.1 | ||||
| U34877.1 | AAC35588.1 | |||
| X93086.1 | CAA63635.1 |
More Information
Set Membership: |
|

Genomic Map