Assay Details
Target Gene Details
Entrez Gene ID: | 168391 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase-like 5 |
Gene Aliases: |
GALNACT19, GALNT15, GalNAc-T5L |
Location: |
Chr.7:151956379-152019934 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNTL5 | NM_145292.3 | NP_660335.2 | ||
| NR_033169.1 | ||||
| XM_006715862.3 | XP_006715925.1 | |||
| XM_017011793.1 | XP_016867282.1 | |||
| XM_017011795.1 | XP_016867284.1 | |||
| XM_017011796.1 | XP_016867285.1 | |||
| AF440400.1 | AAM20912.1 | |||
| AF440404.1 | AAP97318.1 | |||
| AK093091.1 | ||||
| AK315667.1 | ||||
| BC022021.2 | AAH22021.1 | |||
| DB022678.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1029778 | Chr.7:151966767 - 152375920 on Build GRCh38 | Gain |
|
| nsv1020487 | Chr.7:151946824 - 152195828 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map