Assay Details
Target Gene Details
Entrez Gene ID: | 50640 |
Gene Name: | patatin like phospholipase domain containing 8 |
Gene Aliases: |
IPLA2-2, IPLA2G, MMLA, PNPLA-gamma, iPLA2gamma |
Location: |
Chr.7:108470417-108528161 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNPLA8 | NM_001256007.2 | NP_001242936.1 | ||
| NM_001256008.2 | NP_001242937.1 | |||
| NM_001256009.2 | NP_001242938.1 | |||
| NM_001256010.2 | NP_001242939.1 | |||
| NM_001256011.2 | NP_001242940.1 | |||
| NM_015723.4 | NP_056538.1 | |||
| XM_005250396.4 | XP_005250453.1 | |||
| XM_011516274.2 | XP_011514576.1 | |||
| XM_011516275.2 | XP_011514577.1 | |||
| XM_017012261.1 | XP_016867750.1 | |||
| AB041261.1 | BAA94997.1 | |||
| AF217519.1 | AAF67630.1 | |||
| AF263613.1 | AAF75269.1 | |||
| AK002115.1 | BAA92090.1 | |||
| AK024335.1 | BAB14890.1 | |||
| AK291755.1 | ||||
| AL834147.1 | CAD38859.1 | |||
| BC032999.2 | AAH32999.1 | |||
| BX647865.1 |
More Information
Set Membership: |
|

Genomic Map