Assay Details
Target Gene Details
Entrez Gene ID: | 1644 |
Gene Name: | dopa decarboxylase |
Gene Aliases: |
AADC |
Location: |
Chr.7:50458436-50565460 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDC | NM_000790.3 | NP_000781.1 | ||
| NM_001082971.1 | NP_001076440.1 | |||
| NM_001242886.1 | NP_001229815.1 | |||
| NM_001242887.1 | NP_001229816.1 | |||
| NM_001242888.1 | NP_001229817.1 | |||
| NM_001242889.1 | NP_001229818.1 | |||
| NM_001242890.1 | NP_001229819.1 | |||
| XM_005271745.4 | XP_005271802.1 | |||
| AJ310724.1 | CAC84071.1 | |||
| AK298321.1 | ||||
| AK298392.1 | ||||
| AU310260.1 | ||||
| BC000485.1 | AAH00485.1 | |||
| BC008366.1 | AAH08366.1 | |||
| BT007009.1 | AAP35655.1 | |||
| CD013934.1 | ||||
| CD013935.1 | ||||
| CD013936.1 | ||||
| CD013937.1 | ||||
| CD013938.1 | ||||
| CR456724.1 | CAG33005.1 | |||
| DC362073.1 | ||||
| KU178021.1 | ||||
| KU178022.1 | ||||
| M76180.1 | AAA58437.1 | |||
| M88700.1 | AAA20894.1 |
Target Gene Details
Entrez Gene ID: | 100129427 |
Gene Name: | DDC antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:50531759-50543463 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDC-AS1 | NR_033845.1 | 5 | 830 | |
| AK057400.1 | 5 | 830 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016665 | Chr.7:50384854 - 51336827 on Build GRCh38 | Gain |
|
| dgv1286e214 | Chr.7:50485393 - 50585805 on Build GRCh38 | Gain |
|
| nsv1015758 | Chr.7:50497706 - 50700456 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map