Assay Details
Target Gene Details
Entrez Gene ID: | 222194 |
Gene Name: | round spermatid basic protein 1 like |
Gene Aliases: |
- |
Location: |
Chr.7:77696426-77779803 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSBN1L | NM_198467.2 | NP_940869.2 | ||
| BC059402.1 | AAH59402.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs139474842] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map