Assay Details
Target Gene Details
Entrez Gene ID: | 101929269 |
Gene Name: | uncharacterized LOC101929269 |
Gene Aliases: |
- |
Location: |
Chr.8:11062652-11067089 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929269 | NR_134308.1 |
Target Gene Details
Entrez Gene ID: | 286046 |
Gene Name: | XK related 6 |
Gene Aliases: |
C8orf21, C8orf5, C8orf7, XRG6 |
Location: |
Chr.8:10896144-11205789 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XKR6 | NM_173683.3 | NP_775954.2 | ||
| AY534244.1 | AAT07093.1 |

Genomic Map