Assay Details
Target Gene Details
Entrez Gene ID: | 2936 |
Gene Name: | glutathione-disulfide reductase |
Gene Aliases: |
HEL-75, HEL-S-122m |
Location: |
Chr.8:30678061-30727969 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSR | NM_000637.3 | 13 | 2951 | NP_000628.2 |
| NM_001195102.1 | 12 | 2864 | NP_001182031.1 | |
| NM_001195103.1 | 12 | 2792 | NP_001182032.1 | |
| NM_001195104.1 | 11 | 2705 | NP_001182033.1 | |
| CA944703.1 | 1 | 224 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2671289 | Chr.8:30677728 - 30678875 on Build GRCh38 | Deletion |
|
| nsv516544 | Chr.8:30678143 - 30703370 on Build GRCh38 | Loss |
|
| esv3616792 | Chr.8:30677571 - 30678955 on Build GRCh38 | Loss |
|
| esv2657353 | Chr.8:30677311 - 30679064 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map