Assay Details
Target Gene Details
Entrez Gene ID: | 101927221 |
Gene Name: | UBR5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:102239451-102252174 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBR5-AS1 | NR_125415.1 | |||
| AK095151.1 | ||||
| BQ082210.1 | ||||
| DA677213.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576598 | Chr.8:102200959 - 102574604 on Build GRCh38 | Gain |
|
| nsv1025935 | Chr.8:102186979 - 102456094 on Build GRCh38 | Gain |
|
| esv3618218 | Chr.8:102200369 - 102319026 on Build GRCh38 | Gain |
|
| nsv1020359 | Chr.8:102097693 - 102306187 on Build GRCh38 | Gain |
|

Genomic Map