Assay Details
Target Gene Details
Entrez Gene ID: | 29883 |
Gene Name: | CCR4-NOT transcription complex subunit 7 |
Gene Aliases: |
CAF-1, CAF1, Caf1a, hCAF-1 |
Location: |
Chr.8:17226030-17246909 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT7 | XM_005273481.2 | 8 | 2999 | XP_005273538.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831244 | Chr.8:17105917 - 17270710 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map