Assay Details
Target Gene Details
Entrez Gene ID: | 5569 |
Gene Name: | protein kinase (cAMP-dependent, catalytic) inhibitor alpha |
Gene Aliases: |
PRKACN1 |
Location: |
Chr.8:78516101-78605267 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKIA | NM_006823.3 | NP_006814.1 | ||
| NM_181839.2 | NP_862822.1 | |||
| XM_017013620.1 | XP_016869109.1 | |||
| BI597731.1 | ||||
| DB480496.1 | ||||
| S76965.1 | AAB21141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3617735 | Chr.8:78519160 - 78601023 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116951748] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map