Assay Details
Target Gene Details
Entrez Gene ID: | 5569 |
Gene Name: | protein kinase (cAMP-dependent, catalytic) inhibitor alpha |
Gene Aliases: |
PRKACN1 |
Location: |
Chr.8:78516101-78605267 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKIA | NM_006823.3 | NP_006814.1 | ||
| NM_181839.2 | NP_862822.1 | |||
| XM_017013620.1 | XP_016869109.1 | |||
| BI597731.1 | ||||
| DB480496.1 | ||||
| S76965.1 | AAB21141.1 |
Target Gene Details
Entrez Gene ID: | 101927003 |
Gene Name: | PKIA antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:78533892-78558503 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKIA-AS1 | NR_125389.1 | 3 | 917 | |
| BU161278.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3617735 | Chr.8:78519160 - 78601023 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map