Assay Details
Target Gene Details
Entrez Gene ID: | 8767 |
Gene Name: | receptor interacting serine/threonine kinase 2 |
Gene Aliases: |
CARD3, CARDIAK, CCK, GIG30, RICK, RIP2 |
Location: |
Chr.8:89757747-89791064 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIPK2 | NM_003821.5 | NP_003812.1 | ||
| XM_005251092.3 | XP_005251149.1 | |||
| AF027706.1 | AAC34970.1 | |||
| AF064824.1 | AAC25668.1 | |||
| AF078530.1 | AAC27722.1 | |||
| AK075213.1 | ||||
| AK301448.1 | ||||
| AY358813.1 | AAQ89172.1 | |||
| AY358814.1 | AAQ89173.1 | |||
| AY524045.1 | AAS94254.1 | |||
| AY562996.1 | AAS75586.1 | |||
| BC004553.2 | AAH04553.1 | |||
| DA040898.1 | ||||
| KU178437.1 | ||||
| KU178438.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576595 | Chr.8:89459560 - 89771643 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map