Assay Details
Target Gene Details
Entrez Gene ID: | 27085 |
Gene Name: | MDM2 binding protein |
Gene Aliases: |
MDM2BP |
Location: |
Chr.8:120445398-120523635 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTBP | NM_022045.4 | NP_071328.2 | ||
| XM_011516962.2 | XP_011515264.1 | |||
| XM_011516963.2 | XP_011515265.1 | |||
| AK022122.1 | BAB13965.1 | |||
| AK300762.1 | ||||
| BC013136.1 | AAH13136.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519152 | Chr.8:120446217 - 120559026 on Build GRCh38 | Gain |
|
| nsv470235 | Chr.8:120424982 - 120514507 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map