Assay Details
Target Gene Details
Entrez Gene ID: | 2267 |
Gene Name: | fibrinogen like 1 |
Gene Aliases: |
HFREP1, HP-041, LFIRE-1, LFIRE1 |
Location: |
Chr.8:17864380-17909983 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGL1 | NM_004467.3 | NP_004458.3 | ||
| NM_147203.2 | NP_671736.2 | |||
| NM_201552.1 | NP_963846.1 | |||
| NM_201553.1 | NP_963847.1 | |||
| XM_017013232.1 | XP_016868721.1 | |||
| XM_017013233.1 | XP_016868722.1 | |||
| AF042354.1 | ||||
| AF168954.1 | AAM88911.1 | |||
| AK225616.1 | ||||
| AK225619.1 | ||||
| AK289448.1 | ||||
| AL531132.3 | ||||
| AX772838.1 | CAE06268.1 | |||
| BC007047.1 | AAH07047.1 | |||
| BG565379.1 | ||||
| BG566131.1 | ||||
| BG567956.1 | ||||
| BG618053.1 | ||||
| D14446.1 | BAA03336.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831250 | Chr.8:17737238 - 17890332 on Build GRCh38 | Loss |
|
| dgv7121n100 | Chr.8:17766079 - 17957752 on Build GRCh38 | Gain |
|
| nsv1034118 | Chr.8:17876103 - 17916258 on Build GRCh38 | Loss |
|
| nsv610701 | Chr.8:17787519 - 17956578 on Build GRCh38 | Gain |
|
| esv3616503 | Chr.8:17877278 - 17897727 on Build GRCh38 | Gain |
|
| esv3616500 | Chr.8:17771011 - 17959017 on Build GRCh38 | Gain |
|
| esv2762735 | Chr.8:17723646 - 17957764 on Build GRCh38 | Gain |
|
| dgv12044n54 | Chr.8:17868408 - 17896525 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79578967] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map